Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAPPA2
Entrez Gene ID
60676
Gene Name
pappalysin 2
Gene Aliases
PAPP-A2, PAPP-E, PAPPE, PLAC3
Chromosome Location
Chr.1: 176463159 - 176845601 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020318.2 NP_064714.2 8 - 9 4396 68
XM_005245422.3 XP_005245479.1 8 - 9 4443 68
XM_011509857.1 XP_011508159.1 9 - 10 4313 68
XM_011509858.2 XP_011508160.1 8 - 9 4656 68
XM_017002023.1 XP_016857512.1 9 - 10 4494 68
XM_017002024.1 XP_016857513.1 9 - 10 4541 68
XM_017002025.1 XP_016857514.1 8 - 9 4647 68
GenBank mRNA AF311940.1 - 7 - 8 3232 68
AF342989.1 - 8 - 9 4396 68
AJ278348.1 - 7 - 8 3195 68
BC152552.1 - 8 - 9 4384 68

View Full Product Details