The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015562.1 | NP_056377.1 | 6 - 7 | 689 | 143 |
XM_011512670.2 | XP_011510972.1 | 6 - 7 | 637 | 143 | |
XM_011512671.2 | XP_011510973.1 | 5 - 6 | 494 | 143 | |
GenBank mRNA | AB018337.1 | - | 6 - 7 | 617 | 143 |
AK026451.1 | - | 6 - 7 | 642 | 143 | |
AK303630.1 | - | 5 - 6 | 494 | 143 | |
AK307899.1 | - | 2 - 3 | 250 | 143 | |
BC028986.1 | - | 5 - 6 | 521 | 143 | |
BC047420.1 | - | 6 - 7 | 639 | 143 |