The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015562.1 | NP_056377.1 | 8 - 9 | 908 | 83 |
XM_011512670.2 | XP_011510972.1 | 8 - 9 | 856 | 83 | |
XM_011512671.2 | XP_011510973.1 | 7 - 8 | 713 | 83 | |
GenBank mRNA | AB018337.1 | - | 8 - 9 | 836 | 83 |
AK026451.1 | - | 8 - 9 | 861 | 83 | |
AK303630.1 | - | 7 - 8 | 713 | 83 | |
AK307899.1 | - | 4 - 5 | 469 | 83 | |
BC028986.1 | - | 7 - 8 | 740 | 83 |