The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015562.1 | NP_056377.1 | 9 - 10 | 1308 | 96 |
XM_011512670.2 | XP_011510972.1 | 9 - 10 | 1256 | 96 | |
XM_011512671.2 | XP_011510973.1 | 8 - 9 | 1113 | 96 | |
GenBank mRNA | AB018337.1 | - | 9 - 10 | 1236 | 96 |
AK026451.1 | - | 9 - 10 | 1261 | 96 | |
AK303630.1 | - | 8 - 9 | 1113 | 96 | |
AK307899.1 | - | 5 - 6 | 869 | 96 | |
BC028986.1 | - | 8 - 9 | 1140 | 96 | |
BC047420.1 | - | 9 - 10 | 1258 | 96 |