The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286431.1 | NP_001273360.1 | 21 - 22 | 7878 | 100 |
NM_001286432.1 | NP_001273361.1 | 20 - 21 | 7771 | 100 | |
NM_002912.4 | NP_002903.3 | 19 - 20 | 7643 | 100 | |
XM_011536030.2 | XP_011534332.1 | 18 - 19 | 7737 | 100 | |
GenBank mRNA | AB209256.1 | - | 6 - 7 | 1559 | 100 |
AF035537.1 | - | 17 - 18 | 6980 | 100 | |
AF058701.1 | - | 19 - 20 | 7270 | 100 | |
AF071798.1 | - | 18 - 19 | 7508 | 100 | |
AF078695.1 | - | 20 - 21 | 7773 | 100 | |
AF157476.1 | - | 18 - 19 | 7216 | 100 | |
AF179428.1 | - | 19 - 20 | 7344 | 100 | |
AF179429.1 | - | 20 - 21 | 7451 | 100 |