The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286431.1 | NP_001273360.1 | 11 - 12 | 1638 | 114 |
NM_001286432.1 | NP_001273361.1 | 10 - 11 | 1531 | 114 | |
NM_002912.4 | NP_002903.3 | 9 - 10 | 1403 | 114 | |
XM_011536029.2 | XP_011534331.1 | 8 - 9 | 1494 | 114 | |
XM_017011152.1 | XP_016866641.1 | 9 - 10 | 1622 | 114 | |
XM_017011153.1 | XP_016866642.1 | 9 - 10 | 1165 | 114 | |
XM_017011154.1 | XP_016866643.1 | 9 - 10 | 1050 | 114 | |
GenBank mRNA | AF035537.1 | - | 7 - 8 | 740 | 114 |
AF058701.1 | - | 9 - 10 | 1030 | 114 | |
AF071798.1 | - | 8 - 9 | 1268 | 114 | |
AF078695.1 | - | 10 - 11 | 1533 | 114 | |
AF157476.1 | - | 8 - 9 | 976 | 114 | |
AF179428.1 | - | 9 - 10 | 1104 | 114 | |
AF179429.1 | - | 10 - 11 | 1211 | 114 | |
BC035823.1 | - | 8 - 9 | 1202 | 114 |