The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022481.5 | NP_071926.4 | 13 - 14 | 2070 | 59 |
XM_005268497.1 | XP_005268554.1 | 13 - 14 | 2034 | 59 | |
XM_005268498.1 | XP_005268555.1 | 13 - 14 | 2034 | 59 | |
XM_005268499.1 | XP_005268556.1 | 13 - 14 | 2008 | 59 | |
XM_005268500.3 | XP_005268557.1 | 13 - 14 | 2034 | 59 | |
XM_006714792.1 | XP_006714855.1 | 13 - 14 | 2034 | 59 | |
XM_011537677.2 | XP_011535979.1 | 11 - 12 | 1489 | 59 | |
GenBank mRNA | AJ310567.1 | - | 12 - 13 | 2020 | 59 |
AK290052.1 | - | 13 - 14 | 2063 | 59 | |
AK295768.1 | - | 7 - 8 | 1047 | 59 |