The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022481.5 | NP_071926.4 | 8 - 9 | 1352 | 57 |
XM_005268497.1 | XP_005268554.1 | 8 - 9 | 1316 | 57 | |
XM_005268498.1 | XP_005268555.1 | 8 - 9 | 1316 | 57 | |
XM_005268499.1 | XP_005268556.1 | 8 - 9 | 1290 | 57 | |
XM_005268500.3 | XP_005268557.1 | 8 - 9 | 1316 | 57 | |
XM_006714792.1 | XP_006714855.1 | 8 - 9 | 1316 | 57 | |
XM_011537677.2 | XP_011535979.1 | 6 - 7 | 771 | 57 | |
GenBank mRNA | AJ310567.1 | - | 7 - 8 | 1302 | 57 |
AK091385.1 | - | 8 - 9 | 1290 | 57 | |
AK290052.1 | - | 8 - 9 | 1345 | 57 | |
AK295768.1 | - | 2 - 3 | 329 | 57 | |
BC025757.1 | - | 7 - 8 | 1630 | 57 | |
BC027880.1 | - | 7 - 8 | 1619 | 57 |