The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182519.2 | NP_872325.2 | 3 - 4 | 691 | 122 |
GenBank mRNA | AF549190.1 | - | 2 - 3 | 559 | 122 |