Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR36
Entrez Gene ID
134430
Gene Name
WD repeat domain 36
Gene Aliases
GLC1G, TA-WDRP, TAWDRP, UTP21
Chromosome Location
Chr.5: 111092172 - 111130502 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139281.2 NP_644810.1 7 - 8 1017 159
GenBank mRNA AF385437.1 - 7 - 8 1017 159
AK095312.1 - 7 - 8 875 159
AL832494.1 - 6 - 7 823 159
BC133025.1 - 7 - 8 977 159
BC136517.1 - 7 - 8 977 159
CR749211.1 - 2 - 3 217 159

View Full Product Details