The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032604.3 | NP_115993.3 | 8 - 9 | 1167 | 56 |
XM_011533135.2 | XP_011531437.1 | 8 - 9 | 1141 | 56 | |
XM_011533136.2 | XP_011531438.1 | 7 - 8 | 1108 | 56 | |
XM_011533137.2 | XP_011531439.1 | 8 - 9 | 1157 | 56 | |
XM_011533138.2 | XP_011531440.1 | 7 - 8 | 1023 | 56 | |
GenBank mRNA | AK093447.1 | - | 7 - 8 | 1108 | 56 |
AY033290.1 | - | 8 - 9 | 1040 | 56 | |
BC039576.1 | - | 8 - 9 | 1121 | 56 |