Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RER1
Entrez Gene ID
11079
Gene Name
retention in endoplasmic reticulum sorting receptor 1
Gene Aliases
-
Chromosome Location
Chr.1: 2391314 - 2405444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007033.4 NP_008964.3 1 - 2 189 93
GenBank mRNA AF091071.1 - 1 - 2 74 93
AF157324.1 - 1 - 2 92
AK289658.1 - 1 - 2 123 93
BC004965.1 - 1 - 2 124 93

View Full Product Details