The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007033.4 | NP_008964.3 | 4 - 5 | 474 | 128 |
XM_005244713.4 | XP_005244770.2 | 4 - 5 | 1295 | 128 | |
XM_006710306.2 | XP_006710369.1 | 5 - 6 | 556 | 128 | |
XM_011540542.1 | XP_011538844.1 | 5 - 6 | 494 | 128 | |
XM_011540543.1 | XP_011538845.1 | 4 - 5 | 342 | 128 | |
XM_017000131.1 | XP_016855620.1 | 4 - 5 | 1825 | 128 | |
XM_017000132.1 | XP_016855621.1 | 5 - 6 | 584 | 128 | |
XM_017000133.1 | XP_016855622.1 | 5 - 6 | 667 | 128 | |
XM_017000134.1 | XP_016855623.1 | 6 - 7 | 581 | 128 | |
XM_017000135.1 | XP_016855624.1 | 4 - 5 | 581 | 128 | |
GenBank mRNA | AF091071.1 | - | 4 - 5 | 359 | 128 |
AF157324.1 | - | 4 - 5 | 413 | 128 | |
AJ001421.1 | - | 3 - 4 | 283 | 128 | |
AK095750.1 | - | 3 - 4 | 2069 | 128 | |
AK289658.1 | - | 4 - 5 | 408 | 128 | |
BC004965.1 | - | 4 - 5 | 409 | 128 | |
BT007224.1 | - | 3 - 4 | 283 | 128 | |
DA409924.1 | - | 5 - 6 | 556 | 128 |