Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLR3A
Entrez Gene ID
11128
Gene Name
RNA polymerase III subunit A
Gene Aliases
ADDH, HLD7, RPC1, RPC155, hRPC155
Chromosome Location
Chr.10: 77975149 - 78029540 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007055.3 NP_008986.2 29 - 30 4026 62
GenBank mRNA AF021351.1 - 30 - 31 3896 62
AK024744.1 - 1 - 2 84 62
AK026023.1 - 20 - 21 2600 62
AK300329.1 - 30 - 31 3943 62
AY091462.1 - 5 - 6 620 62
BC014399.1 - 6 - 7 732 62
BC041089.1 - 29 - 30 3987 62

View Full Product Details