The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007055.3 | NP_008986.2 | 2 - 3 | 313 | 88 |
GenBank mRNA | AF021351.1 | - | 3 - 4 | 180 | 88 |
AK300329.1 | - | 2 - 3 | 293 | 88 | |
AY091459.1 | - | 2 - 3 | 180 | 88 | |
BC041089.1 | - | 2 - 3 | 274 | 88 | |
DA103416.1 | - | 2 - 3 | 313 | 88 |