The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007055.3 | NP_008986.2 | 5 - 6 | 780 | 122 |
GenBank mRNA | AF021351.1 | - | 6 - 7 | 647 | 122 |
AK300329.1 | - | 5 - 6 | 760 | 122 | |
AY091459.1 | - | 5 - 6 | 647 | 122 | |
BC041089.1 | - | 5 - 6 | 741 | 122 |