Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THOP1
Entrez Gene ID
7064
Gene Name
thimet oligopeptidase 1
Gene Aliases
EP24.15, MEPD_HUMAN, MP78, TOP
Chromosome Location
Chr.19: 2785460 - 2813604 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003249.4 NP_003240.1 6 - 7 955 63
XM_011528228.2 XP_011526530.1 3 - 4 1902 63
GenBank mRNA AK300552.1 - 3 - 4 437 63
BC000135.2 - 6 - 7 881 63
BC000583.2 - 4 - 5 460 63
BC002391.2 - 6 - 7 907 63
BC013878.2 - 6 - 7 871 63
U29366.1 - 6 - 7 839 63
Z50115.1 - 6 - 7 879 63

View Full Product Details