The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001265612.1 | NP_001252541.1 | 11 - 12 | 1546 | 122 |
NM_016284.4 | NP_057368.3 | 11 - 12 | 1546 | 122 | |
NM_206999.2 | NP_996882.1 | 11 - 12 | 1546 | 122 | |
NR_049763.1 | 11 - 12 | 1546 | 122 | ||
GenBank mRNA | AL832769.1 | - | 11 - 12 | 1495 | 122 |
AL833549.1 | - | 11 - 12 | 1506 | 122 | |
BC040523.1 | - | 11 - 12 | 1498 | 122 | |
BX537840.1 | - | 11 - 12 | 1492 | 122 | |
BX647815.1 | - | 11 - 12 | 1486 | 122 | |
CR749237.1 | - | 4 - 5 | 486 | 122 | |
EF553522.1 | - | 11 - 12 | 1495 | 122 |