The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016350.4 | NP_057434.4 | 13 - 14 | 1739 | 111 |
NM_020921.3 | NP_065972.3 | 13 - 14 | 1739 | 111 | |
NM_182944.2 | NP_891989.2 | 13 - 14 | 1739 | 111 | |
NM_182946.1 | NP_891991.1 | 13 - 14 | 1739 | 111 | |
XM_011536817.1 | XP_011535119.1 | 13 - 14 | 1713 | 111 | |
XM_011536819.2 | XP_011535121.1 | 13 - 14 | 1713 | 111 | |
XM_011536822.1 | XP_011535124.1 | 13 - 14 | 1713 | 111 | |
XM_011536823.2 | XP_011535125.1 | 12 - 13 | 1590 | 111 | |
GenBank mRNA | AB046785.2 | - | 13 - 14 | 1664 | 111 |
AF212162.2 | - | 12 - 13 | 1768 | 111 | |
AF302773.2 | - | 13 - 14 | 1739 | 111 | |
AY736182.1 | - | 13 - 14 | 1739 | 111 | |
BC090932.1 | - | 11 - 12 | 1583 | 111 |