The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016350.4 | NP_057434.4 | 23 - 24 | 3350 | 63 |
NM_020921.3 | NP_065972.3 | 24 - 25 | 5489 | 63 | |
NM_182944.2 | NP_891989.2 | 24 - 25 | 5489 | 63 | |
NM_182946.1 | NP_891991.1 | 24 - 25 | 5489 | 63 | |
XM_011536817.1 | XP_011535119.1 | 24 - 25 | 5463 | 63 | |
XM_011536819.2 | XP_011535121.1 | 24 - 25 | 5463 | 63 | |
XM_011536822.1 | XP_011535124.1 | 23 - 24 | 3324 | 63 | |
XM_011536823.2 | XP_011535125.1 | 23 - 24 | 5340 | 63 | |
GenBank mRNA | AB046785.2 | - | 23 - 24 | 3272 | 63 |
AF186776.1 | - | 5 - 6 | 618 | 63 | |
AF212162.2 | - | 23 - 24 | 5518 | 63 | |
AF223937.1 | - | 25 - 26 | 5448 | 63 | |
AF223938.1 | - | 25 - 26 | 5448 | 63 | |
AF223939.1 | - | 25 - 26 | 5448 | 63 | |
AF302773.2 | - | 24 - 25 | 5489 | 63 | |
AK025652.1 | - | 5 - 6 | 609 | 63 | |
AK026083.1 | - | 7 - 8 | 1048 | 63 | |
AY027794.1 | - | 5 - 6 | 618 | 63 | |
AY027795.1 | - | 5 - 6 | 618 | 63 | |
AY027796.1 | - | 5 - 6 | 618 | 63 | |
AY736182.1 | - | 24 - 25 | 5489 | 63 | |
BC090932.1 | - | 21 - 22 | 3194 | 63 |