Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NIN
Entrez Gene ID
51199
Gene Name
ninein
Gene Aliases
SCKL7
Chromosome Location
Chr.14: 50719763 - 50831121 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016350.4 NP_057434.4 9 - 10 1171 94
NM_020921.3 NP_065972.3 9 - 10 1171 94
NM_182944.2 NP_891989.2 9 - 10 1171 94
NM_182946.1 NP_891991.1 9 - 10 1171 94
XM_011536817.1 XP_011535119.1 9 - 10 1145 94
XM_011536819.2 XP_011535121.1 9 - 10 1145 94
XM_011536822.1 XP_011535124.1 9 - 10 1145 94
XM_011536823.2 XP_011535125.1 8 - 9 1022 94
GenBank mRNA AB046785.2 - 9 - 10 1096 94
AF212162.2 - 8 - 9 1200 94
AF223937.1 - 9 - 10 1181 94
AF223938.1 - 9 - 10 1181 94
AF223939.1 - 9 - 10 1181 94
AF302773.2 - 9 - 10 1171 94
AY736182.1 - 9 - 10 1171 94
BC065521.1 - 9 - 10 1081 94
BC090932.1 - 7 - 8 1015 94

View Full Product Details