The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142725.1 | NP_001136197.1 | 2 - 3 | 136 | 90 |
NM_080663.2 | NP_542394.2 | 2 - 3 | 136 | 90 | |
GenBank mRNA | AK127391.1 | - | 2 - 3 | 139 | 90 |
AK292594.1 | - | 3 - 4 | 349 | 90 | |
AK304347.1 | - | 2 - 3 | 135 | 90 | |
BC010503.1 | - | 2 - 3 | 136 | 90 |