The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317962.1 | NP_001304891.1 | 6 - 7 | 637 | 99 |
NM_001317963.1 | NP_001304892.1 | 6 - 7 | 809 | 99 | |
NM_007062.2 | NP_008993.1 | 6 - 7 | 781 | 99 | |
GenBank mRNA | AK223162.1 | - | 6 - 7 | 705 | 99 |
AK290681.1 | - | 6 - 7 | 622 | 99 | |
AK296252.1 | - | 6 - 7 | 638 | 99 | |
AK297961.1 | - | 5 - 6 | 466 | 99 | |
BC000067.2 | - | 6 - 7 | 664 | 99 | |
BC001652.2 | - | 6 - 7 | 700 | 99 | |
BC010921.1 | - | 6 - 7 | 743 | 99 | |
BC032127.2 | - | 6 - 7 | 703 | 99 | |
BC040135.1 | - | 6 - 7 | 679 | 99 | |
BC046144.1 | - | 6 - 7 | 695 | 99 | |
L07758.1 | - | 6 - 7 | 700 | 99 |