The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317962.1 | NP_001304891.1 | 7 - 8 | 770 | 141 |
NM_001317963.1 | NP_001304892.1 | 7 - 8 | 942 | 141 | |
NM_007062.2 | NP_008993.1 | 7 - 8 | 914 | 141 | |
GenBank mRNA | AK223162.1 | - | 7 - 8 | 838 | 141 |
AK290681.1 | - | 7 - 8 | 755 | 141 | |
AK296252.1 | - | 7 - 8 | 771 | 141 | |
AK297961.1 | - | 6 - 7 | 599 | 141 | |
BC001652.2 | - | 7 - 8 | 833 | 141 | |
L07758.1 | - | 7 - 8 | 833 | 141 |