The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190447.1 | NP_001177376.1 | 4 - 5 | 654 | 85 |
NM_002718.4 | NP_002709.2 | 5 - 6 | 3089 | 85 | |
NM_181897.2 | NP_871626.1 | 4 - 5 | 944 | 85 | |
XM_006713686.3 | XP_006713749.1 | 5 - 6 | 3500 | 85 | |
XM_011512956.2 | XP_011511258.1 | 5 - 6 | 3264 | 85 | |
XM_017006787.1 | XP_016862276.1 | 4 - 5 | 2822 | 85 | |
GenBank mRNA | AK293012.1 | - | 5 - 6 | 3069 | 85 |
AK298059.1 | - | 4 - 5 | 583 | 85 | |
AK299544.1 | - | 4 - 5 | 614 | 85 | |
AK302163.1 | - | 3 - 4 | 352 | 85 | |
AK314243.1 | - | 5 - 6 | 3017 | 85 | |
AK316258.1 | - | 4 - 5 | 897 | 85 | |
AL389975.1 | - | 2 - 3 | 172 | 85 | |
BC065531.1 | - | 5 - 6 | 3036 | 85 | |
L07590.1 | - | 5 - 6 | 2976 | 85 | |
L12146.1 | - | 4 - 5 | 869 | 85 |