The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001265612.1 | NP_001252541.1 | 27 - 28 | 4067 | 94 |
NM_016284.4 | NP_057368.3 | 27 - 28 | 4082 | 94 | |
NM_206999.2 | NP_996882.1 | 27 - 28 | 4082 | 94 | |
NR_049763.1 | 27 - 28 | 4067 | 94 | ||
GenBank mRNA | AB023224.2 | - | 14 - 15 | 2127 | 94 |
AK302495.1 | - | 4 - 5 | 452 | 94 | |
AL832769.1 | - | 27 - 28 | 4031 | 94 | |
AL833549.1 | - | 27 - 28 | 4041 | 94 | |
BC024317.1 | - | 10 - 11 | 1481 | 94 | |
BC040523.1 | - | 27 - 28 | 4034 | 94 | |
BX537840.1 | - | 27 - 28 | 4013 | 94 | |
CR749237.1 | - | 20 - 21 | 3022 | 94 | |
EF553522.1 | - | 27 - 28 | 4016 | 94 |