The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204747.1 | NP_001191676.1 | 15 - 16 | 2251 | 70 |
NM_002913.4 | NP_002904.3 | 15 - 16 | 2248 | 70 | |
XM_011513730.1 | XP_011512032.1 | 14 - 15 | 2173 | 70 | |
XM_011513731.1 | XP_011512033.1 | 13 - 14 | 1741 | 70 | |
XM_017008528.1 | XP_016864017.1 | 14 - 15 | 2170 | 70 | |
GenBank mRNA | AF040250.1 | - | 15 - 16 | 2153 | 70 |
AK291612.1 | - | 15 - 16 | 2232 | 70 | |
BC035297.1 | - | 15 - 16 | 2202 | 70 | |
BC051751.1 | - | 15 - 16 | 2218 | 70 | |
BC051786.2 | - | 15 - 16 | 2232 | 70 | |
L23320.1 | - | 15 - 16 | 2317 | 70 |