Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMC2
Entrez Gene ID
9694
Gene Name
ER membrane protein complex subunit 2
Gene Aliases
KIAA0103, TTC35
Chromosome Location
Chr.8: 108443624 - 108486907 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK222615.1 - 1 - 2 62 107
AK290974.1 - 1 - 2 48 107
BC020753.1 - 1 - 2 60 107
BC021667.1 - 1 - 2 77 107
CR457402.1 - 1 - 2 38 107
D14659.1 - 1 - 2 44 107

View Full Product Details