Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMC2
Entrez Gene ID
9694
Gene Name
ER membrane protein complex subunit 2
Gene Aliases
KIAA0103, TTC35
Chromosome Location
Chr.8: 108443624 - 108486907 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK222615.1 - 3 - 4 243 66
AK290974.1 - 3 - 4 229 66
BC020753.1 - 3 - 4 241 66
BC021667.1 - 3 - 4 258 66
CR457402.1 - 3 - 4 219 66
D14659.1 - 3 - 4 225 66

View Full Product Details