The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270987.1 | NP_001257916.1 | 2 - 3 | 327 | 73 |
NM_002342.2 | NP_002333.1 | 2 - 3 | 522 | 73 | |
XM_005253688.1 | XP_005253745.1 | 2 - 3 | 482 | 73 | |
XM_006718983.2 | XP_006719046.1 | 2 - 3 | 482 | 73 | |
GenBank mRNA | AK027080.1 | - | 2 - 3 | 387 | 73 |
BC026262.1 | - | 2 - 3 | 394 | 73 | |
L04270.1 | - | 2 - 3 | 364 | 73 |