The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270987.1 | NP_001257916.1 | 5 - 6 | 701 | 158 |
NM_002342.2 | NP_002333.1 | 5 - 6 | 896 | 158 | |
XM_005253688.1 | XP_005253745.1 | 5 - 6 | 856 | 158 | |
XM_006718983.2 | XP_006719046.1 | 5 - 6 | 856 | 158 | |
GenBank mRNA | AK027080.1 | - | 5 - 6 | 761 | 158 |
AK095418.1 | - | 6 - 7 | 725 | 158 | |
AK293187.1 | - | 5 - 6 | 640 | 158 | |
BC026262.1 | - | 5 - 6 | 768 | 158 | |
L04270.1 | - | 5 - 6 | 738 | 158 | |
L04489.1 | - | 1 - 2 | 347 | 158 |