Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ACOXL
- Entrez Gene ID
-
55289
- Gene Name
- acyl-CoA oxidase-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 110732539 - 111118571 on Build GRCh38
View Full Product Details