The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017408.2 | NP_001017408.1 | 2 - 3 | 702 | 94 |
GenBank mRNA | AF450008.1 | - | 2 - 3 | 690 | 94 |
AK314486.1 | - | 2 - 3 | 673 | 94 | |
AY033606.1 | - | 2 - 3 | 655 | 94 | |
BC009553.2 | - | 2 - 3 | 637 | 94 |