Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL24
Entrez Gene ID
11009
Gene Name
interleukin 24
Gene Aliases
C49A, FISP, IL10B, MDA7, MOB5, ST16
Chromosome Location
Chr.1: 206897404 - 206904139 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185156.1 NP_001172085.1 4 - 5 582 64
NM_006850.3 NP_006841.1 4 - 5 579 64
XM_011509101.2 XP_011507403.1 3 - 4 474 64
XM_017000121.1 XP_016855610.1 3 - 4 472 64
GenBank mRNA AW949784.1 - 2 - 3 214 64
BC009681.1 - 4 - 5 555 64
BT007156.1 - 3 - 4 307 64
U16261.1 - 4 - 5 578 64

View Full Product Details