Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAS1L
Entrez Gene ID
81887
Gene Name
LAS1 like, ribosome biogenesis factor
Gene Aliases
Las1-like, dJ475B7.2
Chromosome Location
Chr.X: 65512582 - 65534806 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170649.1 NP_001164120.1 7 - 8 1051 64
NM_001170650.1 NP_001164121.1 6 - 7 925 64
NM_031206.4 NP_112483.1 7 - 8 1051 64
XM_005262301.1 XP_005262358.1 7 - 8 1036 64
XM_005262304.1 XP_005262361.1 7 - 8 1036 64
XM_005262305.3 XP_005262362.1 7 - 8 1036 64
XM_005262306.3 XP_005262363.1 7 - 8 1036 64
XM_011531045.1 XP_011529347.1 6 - 7 900 64
XM_011531046.1 XP_011529348.1 7 - 8 1036 64
XM_017029877.1 XP_016885366.1 7 - 8 1036 64
XM_017029878.1 XP_016885367.1 7 - 8 1036 64
GenBank mRNA AF116730.1 - 7 - 8 989 64
AK022587.1 - 7 - 8 1001 64
AK097383.1 - 6 - 7 875 64
AK225059.1 - 7 - 8 984 64
BC014545.1 - 7 - 8 1035 64
BC018610.1 - 7 - 8 999 64
BC019302.1 - 7 - 8 996 64

View Full Product Details