Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAT10
Entrez Gene ID
55226
Gene Name
N-acetyltransferase 10
Gene Aliases
ALP, NET43
Chromosome Location
Chr.11: 34105564 - 34146911 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144030.1 NP_001137502.1 15 - 16 1865 63
NM_024662.2 NP_078938.2 17 - 18 2080 63
GenBank mRNA AB051496.1 - 17 - 18 2001 63
AF489535.1 - 13 - 14 1377 63
AK001636.1 - 17 - 18 1935 63
AK022241.1 - 17 - 18 2042 63
AK074430.1 - - 108 63
AK225341.1 - 17 - 18 2042 63
AK294044.1 - 15 - 16 1764 63
AL136882.1 - 17 - 18 1937 63
BC035558.1 - 17 - 18 2014 63

View Full Product Details