Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMPRSS2
Entrez Gene ID
7113
Gene Name
transmembrane protease, serine 2
Gene Aliases
PP9284, PRSS10
Chromosome Location
Chr.21: 41464551 - 41508158 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135099.1 NP_001128571.1 13 - 14 1641 66
NM_005656.3 NP_005647.3 13 - 14 1603 66
XM_005261043.2 XP_005261100.1 12 - 13 1483 66
XM_011529731.2 XP_011528033.1 13 - 14 1698 66
XM_011529733.1 XP_011528035.1 11 - 12 1302 66
GenBank mRNA AF270487.1 - 13 - 14 1582 66
AF329454.1 - 13 - 14 1596 66
AK222784.1 - 12 - 13 1514 66
AK291813.1 - 13 - 14 1558 66
AK296860.1 - 13 - 14 1641 66
BC035623.1 - 13 - 14 1580 66
BC051839.1 - 13 - 14 1574 66

View Full Product Details