The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013291.2 | NP_037423.2 | 13 - 14 | 1371 | 66 |
XM_006716548.2 | XP_006716611.1 | 13 - 14 | 1422 | 66 | |
XM_006716549.2 | XP_006716612.1 | 14 - 15 | 1321 | 66 | |
XM_011516997.1 | XP_011515299.1 | 13 - 14 | 1449 | 66 | |
XM_011516998.1 | XP_011515300.1 | 14 - 15 | 1496 | 66 | |
XM_011516999.1 | XP_011515301.1 | 14 - 15 | 1491 | 66 | |
GenBank mRNA | BC017232.2 | - | 13 - 14 | 1341 | 66 |
U37012.1 | - | 13 - 14 | 1347 | 66 |