The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013291.2 | NP_037423.2 | 3 - 4 | 246 | 78 |
XM_006716548.2 | XP_006716611.1 | 3 - 4 | 297 | 78 | |
GenBank mRNA | AK293601.1 | - | 2 - 3 | 204 | 78 |
BC017232.2 | - | 3 - 4 | 216 | 78 | |
CD299980.1 | - | 3 - 4 | 246 | 78 | |
U37012.1 | - | 3 - 4 | 222 | 78 |