The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135099.1 | NP_001128571.1 | 3 - 4 | 416 | 75 |
NM_005656.3 | NP_005647.3 | 3 - 4 | 378 | 75 | |
XM_005261043.2 | XP_005261100.1 | 3 - 4 | 378 | 75 | |
XM_011529731.2 | XP_011528033.1 | 3 - 4 | 473 | 75 | |
XM_011529732.2 | XP_011528034.1 | 2 - 3 | 300 | 75 | |
GenBank mRNA | AF123453.1 | - | 1 - 2 | 244 | 75 |
AF270487.1 | - | 3 - 4 | 357 | 75 | |
AF329454.1 | - | 3 - 4 | 371 | 75 | |
AK222784.1 | - | 2 - 3 | 289 | 75 | |
AK291813.1 | - | 3 - 4 | 333 | 75 | |
AK296860.1 | - | 3 - 4 | 416 | 75 | |
AK313338.1 | - | 4 - 5 | 678 | 75 | |
BC035623.1 | - | 3 - 4 | 360 | 75 | |
BC051839.1 | - | 3 - 4 | 349 | 75 | |
BP276431.1 | - | 3 - 4 | 378 | 75 | |
U75329.1 | - | 2 - 3 | 300 | 75 |