The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135099.1 | NP_001128571.1 | 8 - 9 | 901 | 71 |
NM_005656.3 | NP_005647.3 | 8 - 9 | 863 | 71 | |
XM_005261043.2 | XP_005261100.1 | 7 - 8 | 743 | 71 | |
XM_011529731.2 | XP_011528033.1 | 8 - 9 | 958 | 71 | |
XM_011529732.2 | XP_011528034.1 | 7 - 8 | 785 | 71 | |
XM_011529733.1 | XP_011528035.1 | 6 - 7 | 562 | 71 | |
GenBank mRNA | AF123453.1 | - | 6 - 7 | 729 | 71 |
AF270487.1 | - | 8 - 9 | 842 | 71 | |
AF329454.1 | - | 8 - 9 | 856 | 71 | |
AK222784.1 | - | 7 - 8 | 774 | 71 | |
AK291813.1 | - | 8 - 9 | 818 | 71 | |
AK296860.1 | - | 8 - 9 | 901 | 71 | |
AK313338.1 | - | 9 - 10 | 1163 | 71 | |
BC035623.1 | - | 8 - 9 | 840 | 71 | |
BC051839.1 | - | 8 - 9 | 834 | 71 |