Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDCD2
Entrez Gene ID
5134
Gene Name
programmed cell death 2
Gene Aliases
RP8, ZMYND7
Chromosome Location
Chr.6: 170575572 - 170584692 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199462.1 NP_001186391.1 5 - 6 734 100
NM_002598.3 NP_002589.2 4 - 5 871 100
XM_017010926.1 XP_016866415.1 4 - 5 812 100
GenBank mRNA AY948416.1 - 5 - 6 661 100
BC008378.1 - 4 - 5 833 100
S78085.1 - 4 - 5 789 100

View Full Product Details