The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164595.1 | NP_001158067.1 | 4 - 5 | 1110 | 102 |
NM_013377.3 | NP_037509.3 | 2 - 3 | 534 | 102 | |
GenBank mRNA | AK091096.1 | - | 2 - 3 | 501 | 102 |
AK131554.1 | - | 4 - 5 | 710 | 102 | |
AK294431.1 | - | 2 - 3 | 590 | 102 | |
AK294535.1 | - | 4 - 5 | 573 | 102 | |
BC093744.1 | - | 2 - 3 | 383 | 102 | |
BC093746.2 | - | 2 - 3 | 383 | 102 |