Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WIZ
Entrez Gene ID
58525
Gene Name
widely interspaced zinc finger motifs
Gene Aliases
ZNF803
Chromosome Location
Chr.19: 15421507 - 15449956 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021241.2 NP_067064.2 1 - 2 156 93
XM_005260004.4 XP_005260061.1 1 - 2 106 93
XM_005260005.4 XP_005260062.1 1 - 2 106 93
XM_005260006.4 XP_005260063.1 1 - 2 106 93
XM_005260007.4 XP_005260064.1 1 - 2 106 93
XM_005260008.2 XP_005260065.1 1 - 2 161 93
XM_005260011.2 XP_005260068.1 1 - 2 161 93
GenBank mRNA AK131404.1 - 1 - 2 157 93

View Full Product Details