The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145031.2 | NP_001138503.1 | 3 - 4 | 621 | 74 |
NM_001319191.1 | NP_001306120.1 | 3 - 4 | 308 | 74 | |
NM_002658.4 | NP_002649.1 | 4 - 5 | 336 | 74 | |
XM_011539866.2 | XP_011538168.1 | 4 - 5 | 331 | 74 | |
GenBank mRNA | A18397.1 | - | 4 - 5 | 286 | 74 |
A35395.1 | - | 4 - 5 | 256 | 74 | |
AK297940.1 | - | 3 - 4 | 277 | 74 | |
AK298560.1 | - | 3 - 4 | 590 | 74 | |
AY029537.1 | - | 3 - 4 | 195 | 74 | |
AY820134.1 | - | 2 - 3 | 127 | 74 | |
BC013575.2 | - | 4 - 5 | 279 | 74 | |
BQ920793.1 | - | 4 - 5 | 329 | 74 | |
BT007391.1 | - | 3 - 4 | 187 | 74 | |
D00244.1 | - | 4 - 5 | 253 | 74 | |
D11143.1 | - | 2 - 3 | 127 | 74 | |
DC349055.1 | - | 4 - 5 | 336 | 74 | |
M15476.1 | - | 4 - 5 | 263 | 74 | |
X02760.1 | - | 3 - 4 | 227 | 74 |