The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145031.2 | NP_001138503.1 | 6 - 7 | 1112 | 62 |
NM_001319191.1 | NP_001306120.1 | 6 - 7 | 799 | 62 | |
NM_002658.4 | NP_002649.1 | 7 - 8 | 827 | 62 | |
XM_011539866.2 | XP_011538168.1 | 7 - 8 | 822 | 62 | |
GenBank mRNA | A18397.1 | - | 7 - 8 | 777 | 62 |
A35395.1 | - | 7 - 8 | 747 | 62 | |
AB208961.1 | - | 1 - 2 | 176 | 62 | |
AK297940.1 | - | 6 - 7 | 768 | 62 | |
AK298560.1 | - | 6 - 7 | 1081 | 62 | |
AK312960.1 | - | 7 - 8 | 796 | 62 | |
AY820134.1 | - | 5 - 6 | 618 | 62 | |
BC013575.2 | - | 7 - 8 | 770 | 62 | |
BT007391.1 | - | 6 - 7 | 678 | 62 | |
D00244.1 | - | 7 - 8 | 744 | 62 | |
D11143.1 | - | 5 - 6 | 618 | 62 | |
M15476.1 | - | 7 - 8 | 754 | 62 | |
X02760.1 | - | 6 - 7 | 718 | 62 |