The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006509.3 | NP_006500.2 | 7 - 8 | 1020 | 75 |
XM_005259127.3 | XP_005259184.1 | 6 - 7 | 990 | 75 | |
XM_005259128.2 | XP_005259185.1 | 7 - 8 | 1020 | 75 | |
GenBank mRNA | AK290594.1 | - | 7 - 8 | 927 | 75 |
BC028013.1 | - | 7 - 8 | 972 | 75 | |
M83221.1 | - | 7 - 8 | 1033 | 75 |