The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007102.2 | NP_001007103.1 | 6 - 7 | 683 | 69 |
NM_032438.2 | NP_115814.1 | 7 - 8 | 758 | 69 | |
XM_005267161.4 | XP_005267218.1 | 7 - 8 | 934 | 69 | |
XM_006715576.3 | XP_006715639.1 | 7 - 8 | 863 | 69 | |
XM_006715578.3 | XP_006715641.1 | 7 - 8 | 742 | 69 | |
XM_011536179.2 | XP_011534481.1 | 7 - 8 | 741 | 69 | |
XM_011536180.2 | XP_011534482.1 | 7 - 8 | 760 | 69 | |
XM_011536181.2 | XP_011534483.1 | 7 - 8 | 695 | 69 | |
XM_011536183.2 | XP_011534485.1 | 7 - 8 | 1291 | 69 | |
XM_011536184.2 | XP_011534486.1 | 7 - 8 | 695 | 69 | |
XM_017011355.1 | XP_016866844.1 | 6 - 7 | 774 | 69 | |
XM_017011356.1 | XP_016866845.1 | 6 - 7 | 666 | 69 | |
GenBank mRNA | AB058701.1 | - | 6 - 7 | 656 | 69 |
AK094137.1 | - | 7 - 8 | 764 | 69 | |
AK290219.1 | - | 6 - 7 | 675 | 69 | |
BC060845.1 | - | 6 - 7 | 677 | 69 |