The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002292.3 | NP_001002292.3 | 10 - 11 | 1604 | 93 |
NM_001193334.1 | NP_001180263.1 | 9 - 10 | 1337 | 93 | |
NM_024911.6 | NP_079187.3 | 10 - 11 | 1665 | 93 | |
XM_011542191.2 | XP_011540493.1 | 10 - 11 | 1588 | 93 | |
XM_011542192.2 | XP_011540494.1 | 10 - 11 | 1598 | 93 | |
XM_017002390.1 | XP_016857879.1 | 10 - 11 | 1432 | 93 | |
GenBank mRNA | AK026744.1 | - | 7 - 8 | 746 | 93 |
AK074984.1 | - | 10 - 11 | 1587 | 93 | |
AK301613.1 | - | 9 - 10 | 1197 | 93 | |
AK309779.1 | - | 11 - 12 | 1544 | 93 | |
AY359035.1 | - | 10 - 11 | 1647 | 93 | |
BC007211.1 | - | 9 - 10 | 1203 | 93 | |
BC110826.1 | - | 10 - 11 | 1566 | 93 | |
BC137109.1 | - | 10 - 11 | 1462 | 93 | |
BC137113.1 | - | 10 - 11 | 1462 | 93 | |
BX538320.1 | - | 10 - 11 | 1604 | 93 | |
BX648748.1 | - | 9 - 10 | 1335 | 93 | |
CA438784.1 | - | 2 - 3 | 93 | ||
DQ323735.1 | - | 10 - 11 | 1634 | 93 |