The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002292.3 | NP_001002292.3 | 7 - 8 | 1309 | 68 |
NM_001193334.1 | NP_001180263.1 | 6 - 7 | 1042 | 68 | |
NM_024911.6 | NP_079187.3 | 7 - 8 | 1370 | 68 | |
XM_011542191.2 | XP_011540493.1 | 7 - 8 | 1293 | 68 | |
XM_011542192.2 | XP_011540494.1 | 7 - 8 | 1303 | 68 | |
XM_017002390.1 | XP_016857879.1 | 7 - 8 | 1137 | 68 | |
GenBank mRNA | AK026744.1 | - | 4 - 5 | 451 | 68 |
AK074984.1 | - | 7 - 8 | 1292 | 68 | |
AK301613.1 | - | 6 - 7 | 902 | 68 | |
AK309779.1 | - | 8 - 9 | 1249 | 68 | |
AY359035.1 | - | 7 - 8 | 1352 | 68 | |
BC007211.1 | - | 6 - 7 | 908 | 68 | |
BC110826.1 | - | 7 - 8 | 1271 | 68 | |
BC137109.1 | - | 7 - 8 | 1167 | 68 | |
BC137113.1 | - | 7 - 8 | 1167 | 68 | |
BX538320.1 | - | 7 - 8 | 1309 | 68 | |
BX648748.1 | - | 6 - 7 | 1040 | 68 | |
DQ323735.1 | - | 7 - 8 | 1339 | 68 |